Learn More
Fisherbrand™ Oval PTFE Stir Bar Product Code.: 16285831
Only undefined left
Add to basket
Fisherbrand™ Oval PTFE Stir Bar
Product Code. 16285831 Shop All Fisherbrand Products
50 mm, 20 mm, Pack of 5
Click to view available options
Length (Metric):
10 mm
15 mm
20 mm
25 mm
30 mm
35 mm
40 mm
50 mm
64 mm
70 mm
Diameter (Metric):
5 mm
6 mm
10 mm
12 mm
13 mm
16 mm
17 mm
20 mm
Unit Size:
Each
Pack of 10
Pack of 5
Product Code. 16285831
Only undefined left
Add to basket
Only undefined left
Add to basket
Provides stable stirring action. Fisherbrand; Oval PTFE Stir Bar povides optimal turbulence for round-bottom flasks and containers.
  • Stable, general-purpose magnetic stir bar
  • Provides excellent turbulence for round-bottom flasks and containers
  • Autoclavable
Specifications
Autoclavable Autoclavable
Certifications/Compliance Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
Color White
Description PTFE encapsulated Alnico V magnet core
For Use With (Equipment) Round bottom flasks
Diameter (Metric) 20 mm
Length (Metric) 50 mm
Quantity 2/Pk.
Shape Oval
Product Content Correction
Product Title
Fisherbrand™ Oval PTFE Stir Bar >