Learn More
Fisherbrand™ Oval PTFE Stir Bar
Shop All Fisherbrand Products
Click to view available options
Length (Metric):
10 mm
15 mm
20 mm
25 mm
30 mm
35 mm
40 mm
50 mm
64 mm
70 mm
Diameter (Metric):
5 mm
6 mm
10 mm
12 mm
13 mm
16 mm
17 mm
20 mm
Unit Size:
Each
Pack of 10
Pack of 5
Specifications
Specifications
| Autoclavable | Autoclavable |
| Certifications/Compliance | Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS) |
| Color | White |
| Description | PTFE encapsulated Alnico V magnet core |
| For Use With (Equipment) | Round bottom flasks |
| Coating | PTFE |
| Diameter (Metric) | 17 mm |
| Length (Metric) | 50 mm |
| Quantity | 2/Pk. |
| Shape | Oval |
| Show More |
Product Content Correction
Product Title
Fisherbrand™ Oval PTFE Stir Bar >