Learn More
Gilac™ Oval Proofing Basket
Oval Proofing Basket ensures that the dough is well ventilated; it can be staggered stacked and nested to save space.
Brand: Gilac™ G070190
This item is not returnable.
View return policy
Description
- The cavity structure ensures that the dough is well ventilated.
- Can be staggered stacked and nested to save space.
- Comes with food contact certificates for optimum food safety and hygiene.
- Easy to clean and machine-washable.
- Light and extremely durable.
- Wide range of 10 product references to meet all needs (0.5 to 2 kg capacity).
- Liner (G072010) available as an option.
Specifications
Proofing Basket | |
1.7 lb. | |
25 x 15 x 9 cm | |
0.75 kg |
Polypropylene | |
0.75 kg | |
Oval |
Product Content Correction
Product Title
Gilac™ Oval Proofing Basket > Capacity: 0.75 kg, Dimensions (L x W x H): 25 x 15 x 9 cm