missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OSCP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12598-1-AP-20UL
This item is not returnable.
View return policy
Description
The function of this protein remains unknown.Specifications
OSCP1 | |
Polyclonal | |
Unconjugated | |
OSCP1 | |
C1orf102, hOSCP1, NOR1, OSCP1, Protein OSCP1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
127700, 230751, 362595 | |
-20°C | |
Liquid |
Western Blot | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q4QQS3, Q8BHW2, Q8WVF1 | |
OSCP1 | |
OSCP1 Fusion Protein Ag3209 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction