Learn More
Thermo Scientific™ Orion™ Universal Power Adapter for Star A Meters
Ensure a constant power supply to your meter with the Thermo Scientific™ Orion™ Star Series Universal Power Adapter for Orion™ Star A series and legacy Orion™ Star series meters
Brand: Thermo Scientific™ 1010003
799.74 NOK valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
Ensure a constant power supply to your meter using a spare or replacement Thermo Scientific™ Orion™ Universal Power Adapter for the Thermo Scientific™ Orion™ Star A series (Star A320, Star A220, Star A210, Star A120 and Star A110 series) benchtop and portable meters.
- Universal 110/2220V 50/60Hz adapter
Specifications
Universal Power Adapter | |
110/220 V, 50/60 Hz | |
Universal power adapter with US, EU, UK and China plug plates |
Meter Accessories | |
50/60 Hz | |
110/220 V |
Product Suggestions
Customers who viewed this item also viewed.
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
New Offer Save Up to 85%