Learn More
Thermo Scientific™ Orion™ Universal Power Adapter for Orion™ Versa Star Meters
Ensure a constant power supply to your meter using a spare or replacement Thermo Scientific™ Orion™ Universal Power Adapter for the Thermo Scientific™ Orion™ Versa Star meters
Brand: Thermo Scientific™ STARA-PWR
672.06 NOK valid until 2025-08-24
Use promo code "24140" to get your promotional price.
Description
Ensure a constant power supply to your meter using a spare or replacement Thermo Scientific™ Orion™ Universal Power Adapter for the Thermo Scientific™ Orion™ Versa Star meters.
Specifications
Orion Versa Star universal power adapter | |
100/240 VAC, 50/60 Hz | |
Universal power adapter with US, EU, UK and China plug plates |
Meter Accessories | |
50/60 Hz | |
100/240 VAC |
Videos
-
Thermo Scientific Orion meters Taking water analysis to the next level
-
Thermo Scientific Orion meters Taking water analysis to the next level
Promotions
-
Multiple Manufacturers
Quality Analysis and Quality Control Testing
Save up to 59% on some of our most popular productsPromo Code: 24140Expires: 2025-08-24
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13