Learn More
Thermo Scientific™ Orion™ Swing Arm Electrode Stand
Quickly move electrodes between samples with the Thermo Scientific™ Orion™ Swing Arm Electrode Stand, featuring 360 degrees of motion and cable management system
Brand: Thermo Scientific™ 090043
Description
Quickly move electrodes between samples with the Thermo Scientific™ Orion™ Swing Arm Electrode Stand, featuring 360 degrees of motion and cable management system for neatness and safety. Includes weighted base, swing arm and electrode holder that accepts up to four sensors and stirrer probe.
- Holds up to five electrodes with 12mm diameters; or four electrodes and one stirrer probe
- Aluminum construction for stability and strength
- 360° motion allows electrodes to be moved vertically, horizontally and front-to-back positioning into beakers or sample containers
- Head holds three electrodes vertically (12mm max. diameter)
- Two additional slots hold electrodes angled 10° (12mm max. diameter)
- 5.5″ diameter weighted base (140mm) with cushioning on the bottom to protect table surfaces
Videos
-
Thermo Scientific Orion meters Taking water analysis to the next level
-
Thermo Scientific Orion meters Taking water analysis to the next level