Learn More
Thermo Scientific™ Orion Star T900 Series Titrator Accessories
Minimize maintenance downtime with easy to install Orion Star T900 series titrator replacement parts. Orion Star T900 series titrator accessories expand functionality.
255.13 NOK - 8300.64 NOK
Specifications
For Use With (Equipment) | Orion Star T910 pH titrator, Orion Star T920 redox titrator, Orion Star T930 ion titrator, Orion Star T940 all-in-one titrator |
---|
Product Code | Brand | Type | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | ||||||
15872107
|
Thermo Scientific™
START-B00 |
Burette Cover |
581.99 NOK
Each |
In Stock
Log in to see stock. |
||||||
15882107
|
Thermo Scientific™
START-B10 |
10 mL Burette |
5043.87 NOK
Each |
Estimated Shipment: 01-07-2025
Log in to see stock. |
||||||
15892107
|
Thermo Scientific™
START-B20 |
20 mL Burette |
5043.87 NOK
Each |
Estimated Shipment: 01-07-2025
Log in to see stock. |
||||||
15802117
|
Thermo Scientific™
START-B50 |
50 mL Burette |
5043.87 NOK
Each |
In Stock
Log in to see stock. |
||||||
15812117
|
Thermo Scientific™
START-BT0 |
Reagent Bottle Holder |
581.99 NOK
Each |
Estimated Shipment: 01-07-2025
Log in to see stock. |
||||||
15822117
|
Thermo Scientific™
START-BT1 |
1 L Plastic Bottle |
330.38 NOK
Each |
In Stock
Log in to see stock. |
||||||
15832117
|
Thermo Scientific™
START-BT2 |
1 L Glass Bottle |
330.38 NOK
Each |
Estimated Shipment: 01-07-2025
Log in to see stock. |
||||||
15842117
|
Thermo Scientific™
START-BT3 |
12 x 1 L Plastic Bottles |
2245.64 NOK
Each |
Estimated Shipment: 01-07-2025
Log in to see stock. |
||||||
15852117
|
Thermo Scientific™
START-BT4 |
12 x 1 L Glass Bottles |
2245.64 NOK
Each |
Estimated Shipment: 01-07-2025
Log in to see stock. |
||||||
15862117
|
Thermo Scientific™
START-CP1 |
Reagent Bottle Cap (GL38) |
701.91 NOK
Each |
Estimated Shipment: 01-07-2025
Log in to see stock. |
||||||
15872117
|
Thermo Scientific™
START-CP2 |
Reagent Bottle Cap (GL45) |
701.91 NOK
Each |
In Stock
Log in to see stock. |
||||||
15882117
|
Thermo Scientific™
START-CP3 |
Pint Bottle Cap |
962.92 NOK
Each |
Estimated Shipment: 01-07-2025
Log in to see stock. |
||||||
15892117
|
Thermo Scientific™
START-DS1 |
Dispenser Probe |
1234.51 NOK
Each |
Estimated Shipment: 01-07-2025
Log in to see stock. |
||||||
15802127
|
Thermo Scientific™
START-DVK |
Dispenser Verification Kit |
581.99 NOK
Each |
Estimated Shipment: 01-07-2025
Log in to see stock. |
||||||
15812127
|
Thermo Scientific™
START-EH1 |
Electrode Holder Assembly |
1234.51 NOK
Each |
Estimated Shipment: 01-07-2025
Log in to see stock. |
||||||
15822127
|
Thermo Scientific™
START-EH2 |
Stopper |
447.95 NOK
Each |
Estimated Shipment: 01-07-2025
Log in to see stock. |
||||||
15832127
|
Thermo Scientific™
START-EH3 |
Cable Management |
255.13 NOK
Each |
Estimated Shipment: 01-07-2025
Log in to see stock. |
||||||
15842127
|
Thermo Scientific™
START-PS2 |
EU Power Supply |
447.95 NOK
Each |
Estimated Shipment: 01-07-2025
Log in to see stock. |
||||||
15852127
|
Thermo Scientific™
START-PS3 |
UK/Singapore Power Supply |
447.95 NOK
Each |
Estimated Shipment: 01-07-2025
Log in to see stock. |
||||||
15862127
|
Thermo Scientific™
START-TB1 |
Standard Tubing Kit |
1422.63 NOK
Each |
Estimated Shipment: 01-07-2025
Log in to see stock. |
||||||
15872127
|
Thermo Scientific™
START-TB2 |
Light-blocking Tubing Kit |
1422.63 NOK
Each |
Estimated Shipment: 01-07-2025
Log in to see stock. |
||||||
15882127
|
Thermo Scientific™
START-TB3 |
Drying Tube |
390.34 NOK
Each |
Estimated Shipment: 01-07-2025
Log in to see stock. |
||||||
15892127
|
Thermo Scientific™
START-TB4 |
Replacemet Burette |
507.92 NOK
Each |
Estimated Shipment: 01-07-2025
Log in to see stock. |
||||||
15802137
|
Thermo Scientific™
START-TB5 |
Replacement Reagent Cap |
507.92 NOK
Each |
Estimated Shipment: 01-07-2025
Log in to see stock. |
||||||
15812137
|
Thermo Scientific™
START-TB6 |
Replacement Dispenser Probe |
507.92 NOK
Each |
Estimated Shipment: 01-07-2025
Log in to see stock. |
||||||
15822137
|
Thermo Scientific™
START-UM1 |
User Manual |
447.95 NOK
Each |
Estimated Shipment: 01-07-2025
Log in to see stock. |
||||||
15832137
|
Thermo Scientific™
START-USB |
USB Cable |
447.95 NOK
Each |
Estimated Shipment: 01-07-2025
Log in to see stock. |
||||||
15842137
|
Thermo Scientific™
START-VK1 |
Value Replacement |
8300.64 NOK
Each |
Estimated Shipment: 01-07-2025
Log in to see stock. |
||||||
Description
Get accessories and replacement parts for the Orion Star T910 pH titrator, Orion Star T920 redox titrator, Orion Star T930 ion titrator and Orion Star T940 all-in-one titrator. These are designed to expand titrator functionality and minimize maintenance downtime.
- Select interchangable burette sizes of 10mL, 20mL or 50mL to customize your titrator
- Bottle holder helps secure titrant to prevent spills and offers flexible placement
- Color-coded tubing and connectors simplify tubing installation and maintenance
Specifications
Orion Star T910 pH titrator, Orion Star T920 redox titrator, Orion Star T930 ion titrator, Orion Star T940 all-in-one titrator |