Learn More
Thermo Scientific™ Orion™ ROSS Ultra™ Glass Bodied pH/ATC Triode™
Click to view available options
Connector Types:
BNC (Waterproof), 8-pin Mini-DIN
BNC (Waterproof), RCA
Unit Size:
Each
Description
Thermo Scientific™ Refillable Orion™ ROSS Ultra™ Triode™ 3-in-1 pH/Automatic Temperature Compensation Electrodes combine the precision and speed of response of the ROSS Ultra™ line with a built-in ATC. Best suited for use in strong acids/bases, pH > 12 or pH < 2, food & cosmetic emulsions, pure water and slurries. Refillable Orion™ ROSS Ultra™ Triodes™ come with a two year warranty.
Measure pH and Temperature with One Electrode
Complete your pH measurements faster. Our ROSS Ultra Triode pH electrodes offer more flexibility, accuracy and precision for your pH measurement needs. With built-in temperature sensors, these electrodes offer a convenient solution for your routine sample measurements. Compatible with Thermo Scientific™ Orion™ Versa Star™, Thermo Scientific™ Orion Star™ A, and Thermo Scientific™ Orion™ Dual Star™ meters, as well as Mettler Toledo™ pH meters.
Target Markets:
Complete your pH measurements faster. Our ROSS Ultra Triode pH electrodes offer more flexibility, accuracy and precision for your pH measurement needs. With built-in temperature sensors, these electrodes offer a convenient solution for your routine sample measurements. Compatible with Thermo Scientific™ Orion™ Versa Star™, Thermo Scientific™ Orion Star™ A, and Thermo Scientific™ Orion™ Dual Star™ meters, as well as Mettler Toledo™ pH meters.
Target Markets:
- Pharmaceutical
- Food Manufacturing
- Academic/Government
- Environmental
- Quality Control
- Research
- General Purpose
Specifications
Specifications
| Description | Orion ROSS Ultra Glass Triode pH/ATC Combination Electrodes with a BNC and RCA Connector and 1 meter cable |
| Connector Types | BNC (Waterproof), RCA |
| Junction | Ceramic |
| Material | Chemical Resistant Glass Body |
| pH Range | 0 to 14 |
| Temperature (Metric) Operating | 0°C to 100°C |
| Type | pH/ATC Combination Electrode |
| Filling Solution Type | ROSS electrode 3 M KCl fill solution (Cat. No. 810007) |
| For Use With (Equipment) | Mettler Toledo Five and Seven series pH meters |
| Internal Reference | ROSS |
| Show More |
Documents
Scientific Resources
Application Notes
Measuring pH in Surface Water
File size: 540.4 kB
Titratable Acidity of Red Wine by Manual Titration (Potentiometric)
File size: 127.4 kB
Measuring pH in Wine and Juice
File size: 155.3 kB
Measuring pH of Concentrated Samples
File size: 416.4 kB
Measuring pH in Low Ionic Strength Solutions
File size: 177.7 kB
Brochures
ROSS pH Electrodes Brochure
File size: 1 MB
Product Information
Manuals
User Manual: Thermo Scientific Orion ROSS Electrodes [EN]
Version: NOV.2016
File size: 509.4 kB
Orion ROSS Ultra, ROSS Ultra Triode, ROSS and PerpHecT ROSS Electrodes
Version: DEC.2016
File size: 509.5 kB
Specification Sheets
Orion ROSS Ultra Triode pH Electrodes Specification Sheet [EN]
File size: 190.4 kB
Product Content Correction
Product Title
Thermo Scientific™ Orion™ ROSS Ultra™ Glass Bodied pH/ATC Triode™ >