Learn More
Thermo Scientific™ Orion™ RDO Replacement Electrode Cap
Use the probe optical cap replacement with RDO probes.
Brand: Thermo Scientific™ 087001
1474.88 NOK valid until 2025-08-24
Use promo code "24140" to get your promotional price.
Description
Use the Thermo Scientific™ Orion™ RDO™ Probe Optical Cap Replacement with Orion RDO probes.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Optical Cap | |
Orion RDO Probes 087010MD, 087020MD, 087030MD, 087050MD, 087100MD | |
Probe Optical Cap |
Promotions
-
Multiple Manufacturers
Quality Analysis and Quality Control Testing
Save up to 59% on some of our most popular productsPromo Code: 24140Expires: 2025-08-24
Do not use solvents or soaps to clean the RDO cap and do not rub the cap with abrasive material. Use of these materials will damage the RDO cap and void the warranty. Do not let salt or mineral deposits form on the RDO cap.