Learn More
Thermo Scientific™ Orion™ pH Electrode Cleaning Solutions
Restore proper electrode performance and prolong electrode life using these pH electrode cleaning solutions.
Brand:  Thermo Scientific™ 911110
 View more versions of this product
Product Code. 11772168
  • 524.38 NOK / Pack of 10
Focus On Clean Water
Use promo code 24139 at checkout
View Available Promotions
Description
Restore proper electrode performance and prolong electrode life using Thermo Scientific™ Orion™ pH Electrode Cleaning Solutions in ready to use bottles.
Cleaning Solutions:
  • Solutions are identified by the letters A, B, C, and D; each letter designates a unique formulation
  • Cleaning solution A is for removing protein contaminants, cleaning solution B is for remov
    TRUSTED_SUSTAINABILITY
Specifications
Specifications
10 Pack of pH Electrode Rinse Solution Pouches
Rinse Solution
Solutions restore proper electrode performance and prolong the useful life of the electrode
10 pk.
pH Electrode
Rinse
Pouch
SDS
Documents Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberDZ1A Certificate TypeCertificate of Analysis Date10/01/2025 Catalog Number911110
Lot NumberCR1A Certificate TypeCertificate of Analysis Date30/12/2024 Catalog Number911110
Lot NumberCO1A Certificate TypeCertificate of Analysis Date27/12/2024 Catalog Number911110
Lot NumberCV1A Certificate TypeCertificate of Analysis Date19/07/2024 Catalog Number911110
Lot NumberCV2A Certificate TypeCertificate of Analysis Date19/07/2024 Catalog Number911110
Promotions
Promotions
  • Multiple Manufacturers
    Shop now for up to 60% off Clean Water Research essentials
    Promo Code: 24139
    Expires2025-08-21
    New Offer
    Save Now
Product Content Correction
Product Title
Thermo Scientific™ Orion™ pH Electrode Cleaning Solutions > Electrode Rinse
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.