Learn More
Thermo Scientific™ Orion™ AQUAfast™ AQ4500 Turbidity Meter Accessories
Ensure your turbidity meter has the accessories it needs, including turbidity vials, primary turbidity standards, Formazin turbidity standards, and more.
Brand:  Thermo Scientific™ AC45ST
3565.86 NOK valid until 2025-08-31
Use promo code "24157" to get your promotional price.
 View more versions of this product
Product Code. 10494362
  • 4009.23 NOK / Each
Spotlight
Use promo code 24157 at checkout
View Available Promotions
Description
Ensure your Thermo Scientific™ Orion™ AQUAfast™ AQ4500 Turbidity Meter has the accessories it needs with our selection of Orion™ AQUAfast™ AQ4500 Turbidity Meter Accessories that include turbidity vials, primary turbidity standards, Formazin turbidity standards, and more. Our Orion AQUAfast AQ4500 turbidity meter has advanced features, including dual light sources and drinking water / wastewater methods approved for U.S. EPA regulatory reporting. Keep pace with changing regulatory protocols; use the White Light LED to comply with EPA 180.1 requirements or Infrared (IR) LED to comply with ISO 7027 requirements. Select from measure modes including EPA 180.1, ISO Nephelometric, ISO Absorbance, % transmittance, EBC and ASBC. This IP67 waterproof meter records 100 data points and offers an average battery life of 2500 hours intermittent use.
Log data into internal memory, or order AQ4CBL cable to send data to printer or download to a computer for easy time and date stamped results storage.
  • U.S. EPA approval for Drinking Water Reporting per 40 CFR Part 141, vol. 74, no. 147, Monday, August 3, 2009. Rules and regulations pg. 38351-38356
  • Dual source to comply with EPA 180.1 and ISO 7027
  • Nephelometric and ratiometric measurements with autoranging
  • IP67 waterproof portable turbidity meter
  • Readings in the range of 0 to 1000 NTU with a choice of units: NT, FTU, FNU, ASBC or EBC
Includes:
  • AQ4500 meter
  • Set of 5 USEPA approved primary calibration standards
  • Seven glass vials
  • User guide
  • Carrying case
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Turbidity Meter Accessory
Primary Standards for AQ4500, SDVB for 0, 1.0, 10, 100, 1000NTU
US EPA approval for Drinking Water Reporting per 40 CFR Part 141, vol. 74, no. 147, Monday, August 3, 2009. Rules and regulations pg 38351-38356.EPA 180.1 and ISO 7031
AQ4500 Turbidimeter
Set of 5 USEPA approved primary calibration standards
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberCW1 Certificate TypeCertificate of Analysis Date10/01/2025 Catalog NumberAC45ST
Lot NumberCR1 Certificate TypeCertificate of Analysis Date31/12/2024 Catalog NumberAC45ST
Lot NumberCS1 Certificate TypeCertificate of Analysis Date30/12/2024 Catalog NumberAC45ST
Lot NumberCQ1 Certificate TypeCertificate of Analysis Date30/12/2024 Catalog NumberAC45ST
Lot NumberCQ2 Certificate TypeCertificate of Analysis Date30/12/2024 Catalog NumberAC45ST
Promotions
Promotions
  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24157
    Expires2025-08-31
    Hot Deal
    *Discount will apply to qualifying orders received by Fisher Scientific between 01/07/2025 and 31/08/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
    Terms & Conditions See Full Promo Details
Product Content Correction
Product Title
Thermo Scientific™ Orion™ AQUAfast™ AQ4500 Turbidity Meter Accessories > Primary standards
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.