Learn More
Thermo Scientific™ Orion™ ROSS Ultra™ Glass Bodied Combination pH Electrode
Capture accurate measurements of a variety of sample types, including solvents, with Orion Glass Body pH Electrodes.
Brand: Thermo Scientific™ 8102BNUWP
4076.83 NOK valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Description
Thermo Scientific Orion ROSS Refillable Electrodes provide optimum accuracy and stability while having the additional advantages of a double junction design. They are available in chemically resistant glass bodies and best suited for use in strong acids/bases, pH > 12 or pH < 2, food & cosmetic emulsions, pure water and slurries. Orion ROSS Ultra Refillable electrodes come with a two-year warranty.
electrode storage solution, 475 ml, pH electrode cleaning solution C, 30 ml and 12 mm electrode
storage bottle
Specifications
Glass body combination electrode | |
Ceramic | |
0 to 14 | |
pH Electrode | |
3M KCl | |
ROSS | |
32°F to 212°F | |
1 m |
WPBNC | |
Glass Body | |
0°C to 100°C | |
<0.002pH/day | |
60ml bottle of Orion ROSS electrode fill solution | |
0.01 | |
39.4 in. |
Product Suggestions
Customers who viewed this item also viewed.
Videos
-
Thermo Scientific Orion meters Taking water analysis to the next level
-
Thermo Scientific Orion meters Taking water analysis to the next level
Special Offers
-
Multiple Manufacturers
The Academic Best Buy Book 2025
Browse exclusive prices for our academic customersPromo Code: 25081Expires: 2025-12-31
-
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24155Expires: 2025-04-30
Days
11Hours
9Minutes
11Seconds
44 -
Multiple Manufacturers
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsPromo Code: 25721Expires: 2025-05-01
New Offer Save Now