Learn More
Organic Electrode Cleaning Solution, Reagecon™
Brand: Reagecon OECS
Quantity | 500 mL |
---|
This item is not returnable.
View return policy
Specifications
500 mL |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Product Content Correction
Product Title
Organic Electrode Cleaning Solution, Reagecon™ > Quantity: 500mL