Learn More
BRAND™ Optical Glass Cavity Slides
With rectangular ground edges.
Brand: BRAND™ 475535
2895.37 NOK valid until 2025-06-13
Use promo code "24123" to get your promotional price.
This item is not returnable.
View return policy
Description
- White (clear) soda-lime glass, hydrolytic class 3
- Single concavity
- Provided in packs of 50
Specifications
50 | |
Soda Lime Glass | |
Optical Glass Slide | |
Clear White |
Ground | |
1.2 to 1.5 mm | |
2 |
Promotions
-
Multiple Manufacturers
Price-Shattering Deals on Lab Glassware
Stock up on lab beakers, flasks and more before these deals disappear.Promo Code: 24123Expires: 2025-06-14
Product Content Correction
Product Title
BRAND™ Optical Glass Cavity Slides > Well Count: 2