Learn More
BRAND™ Optical Glass Cavity Slides
With rectangular ground edges.
Brand:  BRAND™ 475565
3442.43 NOK valid until 2025-06-13
Use promo code "24123" to get your promotional price.
 View more versions of this product
Product Code. 10151391
  • 3940.00 NOK / Pack of 50
Price-Shattering Deals on Lab Glassware
Use promo code 24123 at checkout
View Available Promotions
Only null left
In Stock
Edge
Add to basket
Description
Description
  • White (clear) soda-lime glass, hydrolytic class 3
  • Single concavity
  • Provided in packs of 50
TRUSTED_SUSTAINABILITY
Specifications
Specifications
50
Soda Lime Glass
Optical Glass Slide
Clear White
Ground
1.2 to 1.5 mm
3
Certificates
Promotions
Promotions
Product Content Correction
Product Title
BRAND™ Optical Glass Cavity Slides > Well Count: 3