Learn More
Thermo Scientific™ Open Top Screw Caps for Sample Storage Vials
Open Top Screw Caps for Sample Storage Vials
Brand: Thermo Scientific™ B7807-15
90.31 NOK valid until 2025-04-30
Use promo code "24138" to get your promotional price.
This item is not returnable.
View return policy
Description
Open Top Screw Caps for Sample Storage Vials
Specifications
Black | |
Polypropylene | |
100 |
Open top 15-425 Screw Cap | |
Screw Cap (Open-Top) | |
15 mm |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
-
Multiple Manufacturers
Dairy and Plant-Based Products Testing Promotion
Quality testing products for all your needs.Promo Code: 24134Expires: 2025-03-31
Product Content Correction
Product Title
Thermo Scientific™ Open Top Screw Caps for Sample Storage Vials > Black; 15-425