Learn More
Thermo Scientific™ Open Top Screw Caps for Sample Storage Vials
Open Top Screw Caps for Sample Storage Vials
Brand: Thermo Scientific™ B7807-20
144.71 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Open Top Screw Caps for Sample Storage Vials
Specifications
Black | |
Polypropylene | |
100 |
Open top 20-400 Screw Cap | |
Screw Cap (Open-Top) | |
20 mm |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 7
Product Content Correction
Product Title
Thermo Scientific™ Open Top Screw Caps for Sample Storage Vials > Black; 20-400