Learn More
Köttermann Open Mobile Cabinet
This Open Mobile Cabinet offers versatile storage space with an open-front design.
Brand: Köttermann 315-00092
3743.93 NOK valid until 2024-11-01
Use promo code "21705" to get your promotional price.
This item is not returnable.
View return policy
Description
- Versatile storage space
- Mobile with lockable wheels
- Open-front design provides easy access to stored items
- Many sizes and configurations available
Specifications
White | |
500 x 1200 x 590 mm | |
Epoxy Coated | |
None | |
None | |
Plastic with Brakes | |
Steel |
Mobile | |
None | |
Steel | |
None | |
None | |
Yes |