Learn More
Köttermann One Door Tall Storage Cabinet
Click to view available options
Hinge Side:
Left
Right
No. of Shelves:
3
4
Dimensions (D x W x H):
366 x 600 x 1920 mm
516 x 600 x 1920 mm
Unit Size:
Each
Description
- Tall, slender design expands storage with a small footprint
- Single door protects stored items
Specifications
Specifications
| Color | White |
| Adjustable | No |
| Cabinet Type | Tall Storage |
| Dimensions (D x W x H) | 366 x 600 x 1920 mm |
| Door Type | Hinged |
| Finish | Epoxy Coated |
| For Use With (Application) | Storage |
| Hinge Side | Right |
| Locking Mechanism | Yes |
| No. of Doors | 1 |
| Show More |