Learn More
B Braun™ Omnifix Duo Disposable Syringe
Click to view available options
Volume (Metric):
5 mL
Unit Size:
Pack of 100
Description
- Polypropylene
- Eccentric, Luer slip
- Sterile
- 5mL
Specifications
Specifications
Certifications/Compliance | EN ISO 7886-1 |
Material | Polypropylene |
Product Type | Eccentric, Luer-slip |
Disposable | Single-use |
Quantity | 1/Ea. |
Volume (Metric) | 5 mL |
Sterility | Sterile |
Product Content Correction
Product Title
B Braun™ Omnifix Duo Disposable Syringe >