Learn More
EVOS™ Olympus™ 40X Oil Objective, fluorite, coverslip-corrected
For fluorescence and demanding transmitted-light applications
Brand: EVOS™ AMEP4735
This item is not returnable.
View return policy
Description
- Can be used to view images with different magnification
- Coverslip-corrected objective that is optimized for imaging through #1.5 coverslips
- Offers optical performance from visible light to near infrared light
- Meets needs across spectrum of magnifications and optical specifications
Specifications
Objective Lens | |
EVOS™ FL Auto Imaging System, EVOS™ FL Color Imaging System, EVOS™ FL Imaging System, EVOS™ XL Core Imaging System, EVOS™ XL Imaging System | |
0.2 mm | |
40X | |
1 each |
Olympus | |
Vessels, Slides, Cell Culture Dishes, Flasks, Microtiter Well Plates | |
Fluorite | |
1.30 |
Brochure: Guide to EVOS Objectives
File size: 297.3 kB
Product Content Correction
Product Title
EVOS™ Olympus™ 40X Oil Objective, fluorite, coverslip-corrected > Magnification: 40X; Numerical Aperture: 1.30; Working Distance: 0.2mm