Learn More
Euro Protection™ Okenite Safety Shoes
Euro Protection™ Okenite Safety Shoes are breathable, easy to put on/take off, suitable for food industry use, and feature a composite toe cap for protection.
755.00 NOK - 785.20 NOK
Specifications
Color | White, Gray |
---|---|
Heel Style | Rounded |
Liner Material | Breathable 3D mesh lining |
Material | Microfiber |
Sole Features | Removable, textile and foam insole, injected polyurethane midsole, polyurethane outsole. |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
12014295
|
Euro Protection™
9OKEL37 |
37 |
755.00 NOK
Each |
Estimated Shipment: 03-07-2025
Log in to see stock. |
||||||
12024295
|
Euro Protection™
9OKEL38 |
38 |
755.00 NOK
Each |
Estimated Shipment: 07-07-2025
Log in to see stock. |
||||||
12034295
|
Euro Protection™
9OKEL39 |
39 |
755.00 NOK
Each |
Estimated Shipment: 14-07-2025
Log in to see stock. |
||||||
12054295
|
Euro Protection™
9OKEL41 |
41 |
755.00 NOK
Each |
Estimated Shipment: 09-07-2025
Log in to see stock. |
||||||
12064295
|
Euro Protection™
9OKEL42 |
42 |
755.00 NOK
Each |
Estimated Shipment: 08-07-2025
Log in to see stock. |
||||||
12084295
|
Euro Protection™
9OKEL44 |
44 |
755.00 NOK
Each |
Estimated Shipment: 08-07-2025
Log in to see stock. |
||||||
12094295
|
Euro Protection™
9OKEL45 |
45 |
785.20 NOK
Each |
Estimated Shipment: 09-07-2025
Log in to see stock. |
||||||
Description
- Suitable for food industry use
- Easy-on and easy-off
- Protective cover
- Rounded heel for more comfortable driving
- Microfiber upper
- Breathable 3D mesh lining
- Removable, textile and foam insole
- Composite toe cap
- Injected polyurethane midsole
- Polyurethane outsole
Specifications
White, Gray | |
Breathable 3D mesh lining | |
Removable, textile and foam insole, injected polyurethane midsole, polyurethane outsole. | |
Low-cut | |
427 g per shoe in size 8 | |
Safety Shoe |
Rounded | |
Microfiber | |
Polyurethane | |
Composite | |
EN ISO 20345 : 2011 |
Product Title
Euro Protection™ Okenite Safety Shoes