Learn More
Kern Sohn OIML Class F1 Knob Test Weight
Single test weight constructed of stainless steel.
Brand:  Kern Sohn 327-07
Encompass_Preferred
 View more versions of this product
Product Code. 17332093
  • 653.81 NOK / Each
Only null left
Estimated Shipment: 20-06-2025
Description
  • Knob shape
  • Polished stainless steel
Specifications
Specifications
F1
Stainless Steel
100 g
± 0.5 mg
Certificates
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Kern Sohn OIML Class F1 Knob Test Weight