Learn More
Kern Sohn OIML Class E1 Weight Set, Wooden Box
Weight set constructed of polished stainless steel and housed in a wooden box.
11422.76 NOK - 92181.66 NOK
Specifications
Classification E1
Material Stainless Steel
Case Material Wood
View More Specs
Products 17
Product Code Brand Weight (Metric) Price Quantity & Availability  
Product Code Brand Weight (Metric) Price Quantity & Availability  
17391813
Encompass_Preferred
View Documents
Kern Sohn
303-02
1 mg to 50 g
21589.01 NOK
1 set
View alternative products
Save Up to 
»
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
17311823
Encompass_Preferred
View Documents
Kern Sohn
303-03
1 mg to 100 g
22388.60 NOK
1 set
View alternative products
Save Up to 
»
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
17331823
Encompass_Preferred
View Documents
Kern Sohn
303-04
1 mg to 200 g
26272.34 NOK
1 set
View alternative products
Save Up to 
»
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
17351823
Encompass_Preferred
View Documents
Kern Sohn
303-05
1 mg to 500 g
30270.31 NOK
1 set
View alternative products
Save Up to 
»
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
17371823
Encompass_Preferred
View Documents
Kern Sohn
303-06
1 mg to 1 kg
36552.83 NOK
1 set
View alternative products
Save Up to 
»
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
17391823
Encompass_Preferred
View Documents
Kern Sohn
303-07
1 mg to 2 kg
53458.51 NOK
1 set
View alternative products
Save Up to 
»
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
17311833
Encompass_Preferred
View Documents
Kern Sohn
303-08
1 mg to 5 kg
69221.91 NOK
1 set
View alternative products
Save Up to 
»
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
17331833
Encompass_Preferred
View Documents
Kern Sohn
303-09
1 mg to 10 kg
92181.66 NOK
1 set
View alternative products
Save Up to 
»
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
17351833
Encompass_Preferred
View Documents
Kern Sohn
304-02
1 to 50 g
11422.76 NOK
1 set
View alternative products
Save Up to 
»
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
17371833
Encompass_Preferred
View Documents
Kern Sohn
304-03
1 to 100 g
13478.85 NOK
1 set
View alternative products
Save Up to 
»
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
17391833
Encompass_Preferred
View Documents
Kern Sohn
304-04
1 to 200 g
18504.87 NOK
1 set
View alternative products
Save Up to 
»
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
17311843
Encompass_Preferred
View Documents
Kern Sohn
304-05
1 to 500 g
22160.15 NOK
1 set
View alternative products
Save Up to 
»
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
17331843
Encompass_Preferred
View Documents
Kern Sohn
304-06
1 g to 1 kg
27414.62 NOK
1 set
View alternative products
Save Up to 
»
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
17351843
Encompass_Preferred
View Documents
Kern Sohn
304-07
1 g to 2 kg
43749.17 NOK
1 set
View alternative products
Save Up to 
»
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
17371843
Encompass_Preferred
View Documents
Kern Sohn
304-08
1 g to 5 kg
57456.48 NOK
1 set
View alternative products
Save Up to 
»
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
17391843
Encompass_Preferred
View Documents
Kern Sohn
304-09
1 g to 10 kg
79273.94 NOK
1 set
View alternative products
Save Up to 
»
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
17351873
Encompass_Preferred
View Documents
Kern Sohn
308-42
1 to 500 mg
13707.31 NOK
1 set
View alternative products
Save Up to 
»
Estimated Shipment: 05-12-2024
Description
  • Set of weights
  • Polished stainless steel
  • Milligram weight sets 1 mg to 500 mg are supplied in a removable plastic case
Specifications
Specifications
E1
Stainless Steel
Wood