Learn More
Notrax Oct-O-Mat™ Brushes, Polybag
Oct-O-Mat™ Brush inserts for the Oct-O-Mat provide further scraping action.
145.00 NOK
Product Code | Brand | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | |||||
17722218
|
Notrax
564B0000BL |
Black |
145.00 NOK
Each |
Estimated Shipment: 18-12-2024
Log in to see stock. |
|||||
17732218
|
Notrax
564B0000BU |
Blue |
145.00 NOK
Each |
Estimated Shipment: 18-12-2024
Log in to see stock. |
|||||
17742218
|
Notrax
564B0000GN |
Green |
145.00 NOK
Each |
Estimated Shipment: 18-12-2024
Log in to see stock. |
|||||
17752218
|
Notrax
564B0000GY |
Gray |
145.00 NOK
Each |
Estimated Shipment: 18-12-2024
Log in to see stock. |
|||||
17762218
|
Notrax
564B0000RD |
Red |
145.00 NOK
Each |
Estimated Shipment: 18-12-2024
Log in to see stock. |
|||||
17772218
|
Notrax
564B0000YL |
Yellow |
145.00 NOK
Each |
Estimated Shipment: 18-12-2024
Log in to see stock. |
|||||
Description
- Brush inserts for the Oct-O-Mat provide further scraping action.
- Available in 6 colors.
- Packs of 10.
Specifications
PVC | |
Industrial and Commercial Matting |