Learn More
Motic™ Objective, Achromatic EA 10X/0.25
Designed with chromatic aberration correction for 2 colors. Motic™ Objective, Achromatic EA 10X/0.25 is spherically corrected for one color.
Brand:  Motic™ 12180162
Product Code. 15445102
  • 379.00 NOK / Each
Only null left
Estimated Shipment: 22-08-2025
Add to basket
Description
Description
  • Achromatic.
  • Chromatic aberration correction for 2 colors.
  • Spherical correction for 1 color.
Specifications
Specifications
Achromat Objective
5 mm WD
10X
NA: 0.25
PM Series, DM1802-A, SERIES 18, Series 28 Microscopes
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Motic™ Objective, Achromatic EA 10X/0.25 > Magnification: 10X