missing translation for 'onlineSavingsMsg'
Learn More

OAS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1611.84 NOK - 4593.76 NOK

Specifications

Antigen OAS1
Concentration 0.16 mg/mL
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16803894
View Documents Promotion Details
Proteintech
14955-1-AP-20UL
20 μL
1611.84 NOK
20µL
Please to purchase this item. Need a web account? Register with us today!
16893884
View Documents Promotion Details
Proteintech
14955-1-AP-150UL
150 μL
4593.76 NOK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

OAS1 is an interferon inducible protein that may play a role in mediating resistance to virus infection, control of cell growth, differentiation, and apoptosis. It binds double-stranded RNA and polymerizes ATP into PPP(A2′P5′A)N oligomers, which activate the latent RNase L that, when activated, cleaves single-stranded RNAs. This protein is associated with different subcellular fractions such as mitochondrial, nuclear, and rough/smooth microsomal fractions.
Specifications

Specifications

OAS1
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot
Unconjugated
Rabbit
Human, Rat, Mouse
P00973, P11928, Q05961
192281, 246730, 4938
OAS1 Fusion Protein Ag6793
Primary
-20°C
OAS1
0.16 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
(2 5)oligo(A) synthase 1, 2 5 oligoadenylate synthase 1, 2 5A synthase 1, E18/E16, IFI 4, OAS1, OIAS, OIASI, p46/p42 OAS
OAS1, Oas1a
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications