missing translation for 'onlineSavingsMsg'
Learn More
o-Tolyl isocyanate, 98% Product Code.: 11308705

o-Tolyl isocyanate, 98%

Product Code. 11308705
25 g, 25g
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11308705

Brand: Thermo Scientific Alfa Aesar L12247.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 614-68-6 | C8H7NO | 133.15 g/mol

o-Tolyl isocyanate is used as plastic additives and as heat treatment salt formulations for metals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
o-Tolyl isocyanate is used as plastic additives and as heat treatment salt formulations for metals.

Solubility
Hydrolyzes in water.

Notes
Moisture Sensitive. Incompatible with bases, moisture and oxidizing agents. Store under dry inert gas. Protect from humidity and water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 614-68-6
Molecular Formula C8H7NO
Molecular Weight (g/mol) 133.15
MDL Number MFCD00002009
InChI Key VAYMIYBJLRRIFR-UHFFFAOYSA-N
Synonym o-tolyl isocyanate, 2-methylphenyl isocyanate, 2-tolyl isocyanate, o-methylphenyl isocyanate, o-toluene isocyanate, benzene, 1-isocyanato-2-methyl, 2-methylphenylisocyanate, tolyl isocyanate, isocyanic acid, o-tolyl ester, o-tolylisocyanate
PubChem CID 69194
ChEBI CHEBI:60099
IUPAC Name 1-isocyanato-2-methylbenzene
SMILES CC1=CC=CC=C1N=C=O

Specifications

Density 1.074
Boiling Point 186°C
Flash Point 92°C (197°F)
Odor Pungent
Refractive Index 1.5345
Quantity 25 g
UN Number UN2206
Beilstein 507985
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes in water.
Formula Weight 133.15
Percent Purity 98%
Chemical Name or Material o-Tolyl isocyanate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10254279 Certificate TypeCertificate of Analysis Date17/05/2025 Catalog NumberL12247.14,L12247.06
Lot Number10241935 Certificate TypeCertificate of Analysis Date11/07/2022 Catalog NumberL12247.14,L12247.06
Lot Number10175017 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL12247.14,L12247.06
Lot Number10176601 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL12247.14,L12247.06
Lot Number10199312 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL12247.14,L12247.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
o-Tolyl isocyanate, 98% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.