missing translation for 'onlineSavingsMsg'
Learn More
O-Methylisourea hydrochloride, 98%, Thermo Scientific Chemicals Product Code.: 11417846

O-Methylisourea hydrochloride, 98%, Thermo Scientific Chemicals

Product Code. 11417846
10 g
Click to view available options
Quantity:
10 g
50 g
250 g

Product Code. 11417846

Brand: Thermo Scientific Alfa Aesar A14773.09

Please to purchase this item. Need a web account? Register with us today!

O-methylisourea hydrochloride is used to prepare 1-chloro-2-methyl-isourea. It is also used as an organic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
O-methylisourea hydrochloride is used to prepare 1-chloro-2-methyl-isourea. It is also used as an organic intermediate.

Solubility
Soluble in organic solvents, water, methanol and ethanol.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5329-33-9
Molecular Formula C2H7ClN2O
Molecular Weight (g/mol) 110.541
MDL Number MFCD00035043
InChI Key MUDVUWOLBJRUGF-UHFFFAOYSA-N
Synonym o-methylisourea hydrochloride, methyl carbamimidate hydrochloride, o-methyluronium chloride, 2-methylpseudourea hydrochloride, o-methyl-isourea hydrochloride, methoxymethanimidamide hydrochloride, carbamimidic acid, methyl ester, monohydrochloride, carbamimidic acid, methyl ester, hydrochloride 1:1, 2-methylpseudourea hcl, acmc-20ak09Show MoreShow Less
PubChem CID 3083899
IUPAC Name methyl carbamimidate;hydrochloride
SMILES COC(=N)N.Cl

Specifications

Melting Point ∼116°C (decomposition)
Quantity 10 g
Beilstein 3909619
Solubility Information Soluble in organic solvents,water,methanol and ethanol.
Formula Weight 110.54
Percent Purity 98%
Chemical Name or Material O-Methylisourea hydrochloride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10237679 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA14773.18,A14773.30,A14773.09
Lot NumberJ2226B Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14773.18,A14773.30,A14773.09
Lot Number10142414 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14773.18,A14773.30,A14773.09
Lot Number10220613 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14773.18,A14773.30,A14773.09
Lot Number10126081 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14773.18,A14773.30,A14773.09
compliance-icons
Product Identifier
  • O-Methylisourea hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : O-Methylisourea hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
O-Methylisourea hydrochloride, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.