Learn More
Usbeck Laborgeraete™ Nylon Universal Scissors
129.13 NOK - 165.20 NOK
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | ||||||
10565292
|
Usbeck Laborgeraete™
4280 |
170 mm |
129.13 NOK
Each |
Estimated Shipment: 23-07-2025
Log in to see stock. |
||||||
10075651
|
Usbeck Laborgeraete™
4281 |
230 mm |
165.20 NOK
Each |
Estimated Shipment: 23-07-2025
Log in to see stock. |
||||||
Specifications
Universal Scissors | |
Nylon |
Product Title
Usbeck Laborgeraete™ Nylon Universal Scissors