missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Masterflex™ Nylon Male Pipe Adapter
Designed for quick and easy assembly without tools
280.00 NOK - 324.00 NOK
Specifications
Description | Barb x NPT(M) Connection |
---|---|
Autoclavable | Not Autoclavable |
Material | Nylon |
Sterilization Method | Ethylene Oxide |
Color | Translucent |
Product Code | Brand | Tubing Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Tubing Size | Price | Quantity & Availability | ||||||
11780139
|
Masterflex™
30621-44 |
1/8 in. |
281.00 NOK
Pack of 10 |
Estimated Shipment: 01-07-2025
Log in to see stock. |
||||||
11714959
|
Masterflex™
30621-47 |
1/4 in. |
280.00 NOK
Pack of 10 |
Estimated Shipment: 26-05-2025
Log in to see stock. |
||||||
11704959
|
Masterflex™
30621-63 |
3/8 in. |
324.00 NOK
Pack of 10 |
Estimated Shipment: 26-05-2025
Log in to see stock. |
||||||
Description
- Adapts from a NPT male thread to a hose barb
- Barbed fittings are designed for quick and easy assembly without tools
- Combine these simple, economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30psi)
- Available in packs of ten
Specifications
Barb x NPT(M) Connection | |
Nylon | |
Translucent |
Not Autoclavable | |
Ethylene Oxide | |
Male Pipe Adapter |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Masterflex™ Nylon Male Pipe Adapter