missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Nylon Cloth Printable Labels

Blank, printable nylon cloth labels for Laser Printers.

1430.00 NOK - 8785.00 NOK

Specifications

Description LaserTab Laser Printable Labels
Adhesive Type Permanent Acrylic
Color White
Finish Matte
Material Nylon
View More Specs

Products 2
Product Code Brand Width (Metric) HeightMetric Price Quantity & Availability  
Product Code Brand Width (Metric) HeightMetric Price Quantity & Availability  
11533554
View Documents
Brady™
029680
12.7 mm
1430.00 NOK
1 pack
Estimated Shipment: 21-02-2025
to see stock.
Add to basket
 
10766063
View Documents
Brady™
029674
25.4 mm
8785.00 NOK
1 pack
In Stock
to see stock.
Add to basket
 
Description

Description

  • Blank, printable nylon cloth labels for Laser Printers.
Specifications

Specifications

LaserTab Laser Printable Labels
White
Nylon
Labels
-40°C
-40°F to +194°F
80 lb./in.
0.165 mm
Permanent Acrylic
Matte
Laser
Laser Printers
Rectangle
-40°C to +90°C
0.0065 in.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.