Learn More
Sartorius™ Nutrient Dehydrated Media Pad Sets
Brand: Sartorius™ 14087--47----RDN
2763.16 NOK valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
Chromocult | |
Dehydrated Culture Media | |
Black | |
47 mm |