missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUP54 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16232-1-AP-150UL
This item is not returnable.
View return policy
Description
The nuclear envelope creates distinct nuclear and cytoplasmic compartments in eukaryotic cells. It consists of two concentric membranes perforated by nuclear pores, large protein complexes that form aqueous channels to regulate the flow of macromolecules between the nucleus and the cytoplasm. These complexes are composed of at least 100 different polypeptide subunits, many of which belong to the nucleoporin family. This gene encodes a member of the phe-gly repeat-containing nucleoporin subset.Specifications
NUP54 | |
Polyclonal | |
Unconjugated | |
NUP54 | |
54 kDa nucleoporin, nucleoporin 54kDa, Nucleoporin p54, NUP54 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
269113, 53371, 53372 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P70582, Q7Z3B4, Q8BTS4 | |
Nup54 | |
NUP54 Fusion Protein Ag8882 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction