Learn More
Thermo Scientific™ Nunc™ Polystyrene Roller Bottles
Reliably grow cells with high reproducibility by using Thermo Scientific™ Nunc™ Polystyrene Roller Bottles.
Brand: Thermo Scientific™ 141744
2585.43 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Grow cells with high reproducibility by using highly-reliable Thermo Scientific™ Nunc™ Polystyrene Roller Bottles. Developed for applications such as the industrial scale production of vaccines, monoclonal antibodies and biotherapeutics. Nunc PS Roller Bottles are sterilized by gamma irradiation and meet sterility assurance level (SAL) 10-6.
Features:
- Nunc PS Roller Bottles are sterilized by gamma irradiation and meet sterility assurance level (SAL) 10-6
- One-piece seamless design reduces possibility of leaking through a seam.
- Shallow and deep indents allow for excellent manual and automated handling.
- Proprietary manufacturing process allows for greater impact resistance
- Vertical expanded surfaces version facilitates emptying and reduces retention of product
Recommended for:
- Cell culture expansion
- Vaccine production
- Antibody production
Specifications
Pleated, no indent | |
20 | |
20/Pk., 20/Cs. (Double-bagged) | |
Yes | |
Easy on/off, Vented Cap |
Polystyrene | |
20 | |
1450 cm2 | |
Roller Bottle |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
1373140 | Certificate of Processing | 04/12/2024 | 141744 |
1398609 | Certificate of Processing | 04/12/2024 | 141744 |
1398609 | Certificate of Compliance | 02/02/2024 | 141744 |
1373140 | Certificate of Compliance | 13/12/2022 | 141744 |
1263472 | Certificate of Compliance | 21/09/2019 | 141744 |