Learn More
Thermo Scientific™ Nunc™ Polycarbonate Cell Culture Inserts in Multi-Well Plates
Cultivate most cell types without matrix coating when using Thermo Scientific™ Nunc™ Polycarbonate Membrane Inserts in Multidishes.
Brand: Thermo Scientific™ 140644
2325.00 NOK valid until 2025-12-31
Use promo code "24071" to get your promotional price.
Description
Cultivate most cell types without matrix coating when using Thermo Scientific™ Nunc™ Polycarbonate Membrane Inserts in Multidishes.
Features:
- Thermo Scientific™ Nunclon™ Delta-treated and tested for cell attachment
- Tabs provide better grip and easy handling
- Nontoxic and nonpyrogenic
Ordering Information:
Packed in resealable bags
Specifications
Polycarbonate Cell Culture Inserts with 8 micron pore size in 6-well plate | |
3.14 cm2 | |
1.5 mL | |
8 ÎĽm | |
Multidish 6 | |
24 | |
Clear | |
Sterile |
6 | |
Polycarbonate (Membrane) | |
<0.85 x 105 pores/cm2 | |
Culture Insert | |
6 | |
6-Well Plate | |
Cell culture-treated | |
6 |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
185305 | Certificate of Compliance | 19/03/2025 | 140644 |
183036 | Certificate of Compliance | 02/12/2024 | 140644 |
163931 | Certificate of Compliance | 02/12/2024 | 140644 |
164626 | Certificate of Compliance | 02/12/2024 | 140644 |
183038 | Certificate of Compliance | 02/12/2024 | 140644 |
Promotions
-
Thermo Scientific Nunc
,
Thermo Scientific Nalgene
,
Gibco
,
Invitrogen
,
Thermo Scientific
Cell Biology Promotion
Discover how you can maximise your savings and boost your research to the next level with our top-of-the-line Gibco and Thermo Scientific products.Promo Code: 24071Expires: 2025-12-31
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.