Learn More
Thermo Scientific™ Nunc™ Movette Pipets
Reliably transfer liquid using suction thanks to these movette pipets.
Brand:  Thermo Scientific™ 550970
Additional Details : Weight : 1.90000kg
Product Code. 10000701
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Reliably transfer liquid using suction thanks to Thermo Scientific™ Nunc™ Movette Pipets. Nunc Labware Products are made from high purity resins, and molded using our state-of-the-art processes. Plastic labware is a safer alternative to glass without sacrificing accuracy.
Features:
  • Easy to handle
  • Strong suction capacity
  • Constructed of polyethylene
  • Available in 1.5 and 5mL
Specifications
Specifications
5 mL