Learn More
Thermo Scientific™ Nunc™ MicroWell™ 96-Well, Nunclon Delta-Treated, Flat-Bottom Microplate
Click to view available options
No. of Wells:
96
No. per Pack:
1 Microplate
10 Microplates
No. per Case:
160 Microplates
50 Microplates
Description
Ensure consistent and reliable results using the high quality Thermo Scientific™ Nunc™ F96 MicroWell™ White Polystyrene Plate. White plates are optimized for robotics, plate readers and liquid handling systems and designed for key imaging characteristics.
- Background, autoflorescence and crosstalk are improved when you use Nunc Black or White microplates
- Black microplates are recommended for florescence measurements; with minimum back-scattered light and background fluorescence
- White microplates are recommended for luminescence measurements; with maximum reflection and minimal autoluminescence
- Working volume range: 50 to 250μL/well
- Choice of Thermo Scientific™ Nunclon™ Delta surface for cell-based assays
- Customized bar coding is available
Specifications
Specifications
Barcode | Custom Barcoding Available |
Certifications/Compliance | All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) |
Certified | Nunclon Delta certified for monolayer formation, cloning efficiency, non-cytotoxic, non-pyrogenic, and sterility |
Lid | With Lid |
Array | 8 x 12 |
Packaging | 1/Pk., 50/Cs. |
Product Line | Nunc |
Color | White |
Product Type | Microplate |
Skirt Style | Low Flange Pinchbar |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 15
Documents
Scientific Resources
Application Notes
Technical Bulletin: Nunc FluoroNunc Modules and Plates - The principle behind Q.C. procedure
File size: 159.4 kB
Technical Bulletin: Effects of Enlarged Surface/Volume/Ratio in Solid Phase Assays Documented on the basis of Nunc StarWell Modules
File size: 220.6 kB
Technical Bulletin: Stability of Nunc Immuno MaxiSorp Surfaces
File size: 92.6 kB
Technical Bulletin: Detergent in Polystyrene ELISA
File size: 189.7 kB
Technical Bulletin: Edge effect in Nunc MicroWell ELISA
File size: 181 kB
Brochures
Brochure: Nunc Cell Culture Product Selection Guide
File size: 648.1 kB
Product Brochure: Plate Guide
File size: 2.6 MB
Product Information
Manuals
Technical Data Sheet: Nunc F96 Microwell Plate, PS
Version: August 2015
File size: 136.1 kB
Product Bulletins
The Surface/Volume Ratio in Solid Phase Assays
File size: 570 kB
Warranties
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
185244 | Certificate of Compliance | 08/03/2025 | 136101 |
184073 | Certificate of Compliance | 24/12/2024 | 136101 |
178972 | Certificate of Compliance | 02/12/2024 | 136101 |
185244 | Certificate of Irradiation | 28/02/2025 | 136101 |
184073 | Certificate of Irradiation | 13/12/2024 | 136101 |
Product Content Correction
Product Title
Thermo Scientific™ Nunc™ MicroWell™ 96-Well, Nunclon Delta-Treated, Flat-Bottom Microplate >