Learn More
Thermo Scientific™ Nunc™ MicroWell™ 96-Well Microplates
Obtain consistent results with Thermo Scientific™ Nunc™ MicroWell™ 96-Well Microplates, optimized for robotics, automated readers and liquid handling systems.
4380.00 NOK - 4925.00 NOK
Specifications
Barcode | No Barcode |
---|---|
Certifications/Compliance | All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) |
Format | Standard |
Lid | With Lid |
Array | 8 x 12 |
Product Code | Brand | No. per Case | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. per Case | Price | Quantity & Availability | ||||||
10774957
![]() |
Thermo Scientific™
243656 |
108 |
4380.00 NOK
Pack of 108 |
In Stock
Log in to see stock. |
||||||
10000571
![]() |
Thermo Scientific™
260860 |
160 |
4925.00 NOK
Pack of 160 |
In Stock
Log in to see stock. |
||||||
Description
Obtain consistent and reliable results with Thermo Scientific™ Nunc™ MicroWell™ 96-Well Microplates. With multiple surface modifications, these plates are ideal for cell-based assays as well as biochemical assays. The flat bottom shape is suitable for microscopic and optical measurements. Available in either the high flange skirt type or the low flange pinchbar design, the plates are optimized for a range of robotics, automated readers and liquid handling systems.
- Made from clear polystyrene with excellent optical properties, ideal for optical measurements.
- Non-treated plate with hydrophobic surface is suited for suspension cell cultures and a variety of biochemical assays.
- Cell culture plate received the Thermo Scientific™ Nunclon™ Delta surface treatment that promotes cell adhesion and growth
- Collagen I and Poly-D-Lysine coated plates are enhanced for finicky adherent cells and biomolecule attachment
- Different flange designs are compatible with either manual or robotic operations
- Customized bar coding is available
Specifications
No Barcode | |
Standard | |
8 x 12 | |
Clear | |
Polystyrene | |
Sterile | |
Flat Bottom (F-well) |
All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) | |
With Lid | |
Nunc | |
Microplate | |
96 | |
Non-treated |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.