Learn More
Thermo Scientific™ Nunc™ Cell Culture Treated Flasks with Solid Caps
Achieve maximum adhesion for a broad range of cell types.
Brand: Thermo Scientific™ 163371
Description
Achieve maximum adhesion for a broad range of cell types with Thermo Scientific™ Nunc™ Cell Culture Treated Flasks with Solid Caps. These flasks receive the proprietary Nunclon™ Delta surface treatment to ensure consistent cell growth. The vent/close caps with Y marks allow easy visual verification of venting position while promoting sufficient gas exchange.
Features:
- Nunclon Delta Certification including testing on four different cell types is provided in each case.
- Short, wide necks allow for easy access.
- Sterilized by gamma irradiation and certified to be non-pyrogenic.
Ordering Information:
Extra caps included in every carton.
Includes:
Caps
Specifications
Nunc Flask 25 cm², Solid Cap, Angled Neck, 160/Cs | |
Angled | |
20 | |
Flat | |
25 cm² |
Polystyrene | |
160 | |
Sterile | |
Solid | |
Nunclon Delta |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
167518 | Certificate of Compliance | 06/12/2024 | 163371 |
164988 | Certificate of Compliance | 06/12/2024 | 163371 |
159683 | Certificate of Compliance | 06/12/2024 | 163371 |
158550 | Certificate of Compliance | 03/12/2024 | 163371 |
168943 | Certificate of Compliance | 03/12/2024 | 163371 |
Accessories and Services
-
Thermo Scientific™ Caps for Nunclon™ Delta Flasks X100 CAP 25CM² V/C vent/close, sterile Product Code: 10154271 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.