Learn More
Thermo Scientific™ Nunc™ 96-Well, Nunclon Delta-Treated, U-Shaped-Bottom Microplate
Color:Clear; Sterility:Sterile; Lid:With Lid
Brand: Thermo Scientific™ 168136
4830.24 NOK valid until 2025-03-29
Use promo code "24111" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
Thermo Scientific™ Nunc 96-Well Polystyrene Round Bottom Microwell Plates are optimized for robotics, automated readers and liquid handling systems. 96-well round bottom (U) microplates are ideal for agglutination assays.
- Clear polystyrene has excellent optical properties making it ideal for optical measurements
- Untreated polystyrene is hydrophobic and is suited for growth of suspension cultures that can proliferate and grow without attachment
- Untreated surfaces are ideal for adding coatings in your lab
- Cell Culture Treated: The Nunclon™ Delta cell culture treated surface is a hydrophilic surface that promotes cell attachment and growth
- Plates are optimized for robotics, plate readers and liquid handlers
- Customized bar coding is available
Specifications
Custom Barcoding Available | |
Nunclon Delta certified for monolayer formation, cloning efficiency, non-cytotoxic, non-pyrogenic, and sterility | |
8 x 12 | |
Nunc | |
Microplate | |
300 μL | |
Sterile | |
Round Bottom (U-well) | |
10 |
All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) | |
With Lid | |
10 Pk., 160 Case | |
Clear | |
Polystyrene | |
96 | |
Nunclon Delta | |
160 |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
184804 | Certificate of Irradiation | 07/02/2025 | 168136 |
184090 | Certificate of Irradiation | 14/12/2024 | 168136 |
184804 | Certificate of Compliance | 22/01/2025 | 168136 |
184090 | Certificate of Compliance | 10/12/2024 | 168136 |
164627 | Certificate of Compliance | 06/12/2024 | 168136 |
Special Offers
- Multiple Manufacturers
-
Multiple Manufacturers
New Offer Save Now