Learn More
Thermo Scientific™ 384 Well Plate, Non-Treated Surface, No Lid, Non-Sterile, Pack of 25
Color:Clear; Sterility:Non-sterile
Brand: Thermo Scientific™ 262160
4880.24 NOK valid until 2025-03-29
Use promo code "24111" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
Thermo Scientific™ Nunc 384-Well Clear Polystyrene Plates with Cell Culture Treated and Nontreated Surfaces are optimized for robotics, plate readers and liquid handling systems. 384-well microplates allow sample and reagent volume reduction providing cost savings and allowing more samples to be screened. Our plates are high quality for consistent and reliable results.
Nunc 384-well polystyrene plates have rounded square-well geometry, eliminating wicking.- Clear Polystyrene has excellent optical properties making it ideal for optical measurements
- Untreated Polystyrene is hydrophobic and is suited for growth of suspension cultures that can proliferate and grow without attachment
- Untreated surfaces are ideal for adding coatings in your lab
- Cell Culture Treated: The Nunclon Delta cell culture treated surface is a hydrophilic surface that promotes cell attachment and growth
- Customized bar coding is available
- Flexible working volumes; 10-100μL
Specifications
Custom Barcoding Available | |
Without Lid | |
Non-Treated; no lid; Non-sterile; 25/100 | |
25/Pk., 100/Cs. | |
Clear | |
Polystyrene | |
384 | |
Non-sterile | |
Flat Bottom (F-well) | |
25 |
All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) | |
0.084 cm2/well | |
16 x 24 | |
Nunc | |
Microplate | |
120 μL | |
10 to 100 μL | |
Non-treated | |
100 |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
185442 | Certificate of Compliance | 11/03/2025 | 262160 |
184870 | Certificate of Compliance | 15/01/2025 | 262160 |
1420381 | Certificate of Compliance | 14/01/2025 | 262160 |
1313223 | Certificate of Compliance | 11/12/2024 | 262160 |
179018 | Certificate of Compliance | 06/12/2024 | 262160 |
Special Offers
-
Multiple Manufacturers
Grow Your Discovery
Fall in love with big savingsPromo Code: 24107Expires: 2025-03-28
Days
5Hours
15Minutes
24Seconds
56