Learn More
Cytiva Nuclepore Membrane
Brand:  Cytiva 800203
Product Code. 12495412
  • 2300.00 NOK / Each
Only null left
Estimated Shipment: 19-03-2029
Specifications
15 μm
1 Each
Nucleopore Membrane
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Cytiva Nuclepore Membrane > Pore Size: 15μm