Learn More
Tocris Bioscience™ NTRC 824
Potent and selective non-peptide NTS2 antagonist
Brand: Tocris Bioscience™ 5438/50
This item is not returnable.
View return policy
Description
NTRC 824 is a potent and selective non-peptide neurotensin (NTS) type-2 receptor antagonist (IC50 = 38 nM). Exhibits >150-fold selectivity for NTS2 over NTS1.
Specifications
N-[2-[5-[[(4-Methylphenyl)sulfonyl]amino]-3-(2,2,2-trifluoroacetyl)-1H-indol-1-yl]acetyl]-L-leucine | |
50 mg | |
C25H26F3N3O6S | |
>98% |
1623002-61-8 | |
Neurotensin Receptor Antagonists | |
553.55 |
Product Content Correction
Product Title
Tocris Bioscience™ NTRC 824 > 50mg