missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ NTNG1 Recombinant Protein
Brand: Abnova™ H00022854-P01.25ug
This item is not returnable.
View return policy
Description
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
AAH30220 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
70.84 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
HYDLCKTQIYTEEGKVWDYMACQPESTDMTKYLKVKLDPPDITCGDPPETFCAMGNPYMCNNECDASTPELAHPPELMFDFEGRHPSTFWQSATWKEYPKPLQVNITLSWSKTIELTDNIVITFESGRPDQMILEKSLDYGRTWQPYQYYATDCLDAFHMDPKSVKDLSQHTVLEIICTEEYSTGYTTNSKIIHFEIKDRFAFFAGPRLRNMASLYGQLDTTKKLRDFFTVTDLRIRLLRPAVGEIFVDELHLARYFYAISDIKVRGRCKCNLHATVCVYDNSKLTCECEHNTTGPDCGKCKKNYQGRPWSPGSYLPIPKGTANTCIPSISSIGTNVCDNELLHCQNGGTCHNNVRCLCPAAYTGILCEKLRCEEAGSCGSDSGQGAPPHGSPALLLLTTLLGTASPLVF | |
KIAA0976/Lmnt1 | |
NTNG1 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Protein Array, ELISA, Western Blot | |
22854 | |
NTNG1 (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
NTNG1 | |
Human | |
Recombinant | |
Solution |