Learn More
NP-40 lysis buffer (2X) Product Code.: 15415109
NP-40 lysis buffer (2X)
Product Code. 15415109
125 mL, 125mL
Click to view available options
Quantity:
125 mL
250 mL
Unit Size:
125mL
250mL
Product Code. 15415109
Brand: Thermo Scientific Alfa Aesar J62805.=E
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Color Colorless
Concentration 2X
Content And Storage Ambient temperatures
Format Liquid
Quantity 125 mL
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Product Code
Lot NumberR01L553 Certificate TypeCertificate of Analysis Date17/05/2025 Product CodeJ62805.=E
Lot NumberP29L508 Certificate TypeCertificate of Analysis Date17/05/2025 Product CodeJ62805.=E
Lot NumberW16K542 Certificate TypeCertificate of Analysis Date24/09/2024 Product CodeJ62805.=E, J62805.AK
Lot NumberP24K508 Certificate TypeCertificate of Analysis Date11/06/2024 Product CodeJ62805.=E, J62805.AK
Lot NumberZ06J506 Certificate TypeCertificate of Analysis Date24/01/2024 Product CodeJ62805.=E, J62805.AK
Product Title
NP-40 lysis buffer (2X) >