Learn More
Novobiocin Sodium Salt, MP Biomedicals™
Inhibits DNA synthesis by inhibiting the enzyme Topoisomerase II. Novobiocin Sodium Salt, MP Biomedicals is an antibiotic produced by Streptomyces nieveus. It is active against gram-positive bacteria. It is also a Hsp90 inhibitor 3.
Brand: MP Biomedicals 0215595701_CLEAN
Description
Inhibits DNA synthesis by inhibiting the enzyme Topoisomerase II. Novobiocin Sodium Salt, MP Biomedicals is an antibiotic produced by Streptomyces nieveus. It is active against gram-positive bacteria. It is also a Hsp90 inhibitor 3.
Novobiocin inhibits DNA synthesis by inhibiting bacterial DNA gyrase targeting the GyrB subunit of the enzyme involved in energy transduction. It acts as a competitive inhibitor of the ATPase reaction catalysed by GyrB 2. Uses include:
- For the production of positively supercoiled plasmid DNA
- Inhibitor of bacterial DNA gyrase and eukaryotic DNA topoisomerase
- Inhibitor of retrovirus RNA-dependent DNA-polymerase
- To study heat shock protein inhibition