Learn More
Invitrogen™ Colloidal Blue Staining Kit Product Code.: 10593043
Invitrogen™ Colloidal Blue Staining Kit
Product Code. 10593043
1 kit, 0.5 L
Click to view available options
Quantity:
1 kit
Unit Size:
0.5 L
Product Code. 10593043
Brand: Invitrogen™ LC6025
Edge
Includes
The Colloidal Blue Staining Kit is supplied with sufficient stainer A and stainer B to stain 25 mini-gels
Five times more sensitivity than Coomassie™ R-250— and can detect as little as 10ng of BSA
The Colloidal Blue Staining Kit allows detection of nanogram levels of protein with minimal effort and requires only water to destain. The Colloidal Blue stain uses colloidal chemistry that reduces free dye in solution and improves the protein-to-dye binding ratio. Samples are intensely stained and visible within 3 hours, background staining is virtually eliminated by destaining overnight with water.
  • Easy-to-use, mass spectrometry compatible reagents
  • Great densitometry results
Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Room Temperature
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage The Colloidal Blue Staining Kit is supplied with sufficient stainer A and stainer B to stain 25 mini-gels. Store the kit at room temperature. The kit is guaranteed for 6 months when properly stored unless otherwise stated in the product documentation.
Detection Location In-Gel Detection
Detection Method Colorimetric
Label or Dye Coomassie
Product Type Protein Gel Staining Kit
Quantity 1 kit
Shipping Condition Room Temperature
Target Molecule Protein
Manuals
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Product Code
Lot Number3065270 Certificate TypeCertificate of Analysis Date31/12/2024 Product CodeLC6025
Lot Number2942098 Certificate TypeCertificate of Analysis Date10/10/2024 Product CodeLC6025
Lot Number2865812 Certificate TypeCertificate of Analysis Date09/04/2024 Product CodeLC6025
Lot Number2801400 Certificate TypeCertificate of Analysis Date08/01/2024 Product CodeLC6025
Lot Number2740256 Certificate TypeCertificate of Analysis Date06/11/2023 Product CodeLC6025
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Product Title
Invitrogen™ Colloidal Blue Staining Kit >