Learn More
GPE Scientific™ Norell™ Coaxial Inserts For NMR Sample Tubes
Brand:  GPE Scientific™ NI5CCI-B
 View more versions of this product
Product Code. 12812316
  • 1310.00 NOK / Each
Only null left
Estimated Shipment: 14-08-2025
Specifications
Cap for the insert tube and special cap to seal the insert to the top of the outer NMR tube
Coaxial Insert
206 mm
100μL
Certificates
Promotions
Promotions
Product Content Correction
Product Title
GPE Scientific™ Norell™ Coaxial Inserts For NMR Sample Tubes > Capacity: 100μL